Anti-PUM2 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-387
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Pumilio 2
- Antigen symbol:PUM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:23369
- Antigen synonyms:KIAA0235PUMH2PUML2|pumilio homolog 2|MGC138253|FLJ36528|MGC138251|pumilio (Drosphila) homolog 2|Pumilio-2|pumilio homolog 2 (Drosophila)
- Storage buffer:Tris-Citrate/phosphate (pH 7 - 8)
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:1 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Pumilio 2 using the numbering given in entry NP_056132.1 (GeneID 23369)
- Purification:Immunogen affinity purified
- Pk:100 µl
Specifications
About this item
The PUM2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PUM2. This antibody reacts with human, mouse, rat. The PUM2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
There is one scientific publication using this antibody.
Type: Primary
Antigen: PUM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat