Orders LinkContactUsLinkComponent
VWR® Viscometers
VWR® Viscometers
Supplier:   VWR Collection
VWR® Viscometers
Supplier:  VWR Collection
Certificates
VWR® Viscometers
VWR® Viscometers
2 Options Available Below
Product Details & Documents
The Cannon-Fenske routine viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104.
Minimum sample volume is 11 ml. Minimum bath depth is 241 mm (9,5")
Product Family Options
Product Information
  • Range
    Constant
    Availability
    Price
  • VWRU10791-456
    Calibrated, size: 0C
    Calibrated, size: 0C
    0,6 to 3 cSt
    0,003 cSt/s K
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 432,00 €
    432,00 €
    1 * 1 items
       432,00 €
  • VWRU10791-458
    Calibrated, size: 0B
    Calibrated, size: 0B
    1 to 5 cSt
    0,005 cSt/s K
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 432,00 €
    432,00 €
    1 * 1 items
       432,00 €