Orders LinkContactUsLinkComponent
Wire Baskets for Test Tubes
Wire Baskets for Test Tubes
Wire Baskets for Test Tubes
Certificates
Wire Baskets for Test Tubes
3 Options Available Below
Product Details & Documents
Wire basket of welded mesh w 8 mm, d 1 mm for test tubes and more. Made up of stainless steel W-no. 1.4301, cube shape, electrochemically polished.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 100×100×100 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 120×120×120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 34,53 €
    34,53 €
    1 * 1 items
       34,53 €
  • 140×140×140 mm