Accessories for ductless fume hoods, New captair® flex™
Supplier: ERLAB CAPTAIRE
Certificates
About this item
Frequently Bought Together
(±)-1-Methoxy-2-propanol ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Gas washing bottles
Supplier: Lenz Laborglas GmbH & CO.KG
Adapters with screw thread and standard gound joint cone
Supplier: Lenz Laborglas GmbH & CO.KG
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Jars, wide mouth, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Screw caps, bakelite
Supplier: GRAVIS
VWR®, Bottles, HDPE
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
FILTRE MF4BE+ 1 * 1 items
Supplier: ERLAB CAPTAIRE
Accessories for ductless fume hoods, New captair® flex™
Supplier: ERLAB CAPTAIRE
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
Alcometer fuel cell breathalyser
Supplier: SURESCREEN
PREFILTER PARTICULATE FOR 714 SMART 1 * 1 items
Supplier: ERLAB CAPTAIRE
Spare valve plug, 10 mm, PTFE, old model 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Micro-fluidic platform, SilFlow™
Supplier: Trajan Scientific and Medical
α-Methylstyrene 99% stabilised
Supplier: Thermo Fisher Scientific
Test sieves
Supplier: Retsch GmbH