Orders LinkContactUsLinkComponent
TAE buffer, 50X
TAE buffer, 50X
TAE Buffer (50X), Glass bottle, 100 mL
Catalog # EDVO607
Supplier:   EDVOTEK
TAE buffer, 50X
Catalog # EDVO607
Supplier:  EDVOTEK
CAS Number:  
TAE buffer, 50X
TAE Buffer (50X), Glass bottle, 100 mL
CAS Number:  
Price
33,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
This 50X concentrated solution (Tris-acetate, EDTA, pH 7,8) is sufficient for making 5 L of diluted working buffer for making agarose gels and electrophoresis chamber buffer.
Specifications
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    TAE Buffer (50X)
Specifications