Orders LinkContactUsLinkComponent
3M™ Squeegee, White
3M™ Squeegee, White
3M™ squeegee, White, 100×70×10 mm, Felt
Catalog # KROS31612
Supplier:   KROSCHKE
3M™ Squeegee, White
Catalog # KROS31612
Supplier:  KROSCHKE
3M™ Squeegee, White
3M™ squeegee, White, 100×70×10 mm, Felt
Price
32,50 €
Log in for availability
1 * 1 items
Product Details & Documents
3M squeegee, for attaching sensitive films, well suited for detailed work, for profiled surfaces, particularly adaptable, for wet and dry bonding.
Specifications
  • Colour:
    White
  • Description:
    3M™ squeegee
  • Dimensions:
    100×70×10 mm
  • Material:
    Felt