ContactUsLinkComponent
Magic LAB® Homogeniser with DR 2000/03 Module
Magic LAB® Homogeniser with DR 2000/03 Module
Catalog # IKAIMAGICLAB
Supplier:  IKA
Magic LAB® Homogeniser with DR 2000/03 Module
Catalog # IKAIMAGICLAB
Supplier:  IKA
Specifications
  • Description:
    Magic LAB® with module DR, Maximum flow rate: 80 L/hr, Motor power: 0,9 kW, Circumferential speed: 23 m/s, Motor speed: 3000 min⁻¹
  • Pk:
    1
Specifications
About this item
The magic LAB® is a unique and multi-functional small-scale laboratory machine. It is designed for mixing, dispersing, wet milling and incorporation of powders into liquids. The magic LAB® is most frequently used for the development of new products or for optimizing of existing process techniques, particularly in the chemical, cosmetic, pharmaceutical and food industries.
  • Depending on the application, the basic unit can be modified quickly with different working modules
  • Flexibility and ease of use: one machine suits for many applications and variety of processes
  • Simple heating or cooling of all modules
  • User-friendly operation via the magic LAB® controller
  • Practical transport and storage packaging
It is an ideal machine for continuous, circulating and batch processing with interchangeable modules. Many work parameters of the magic LAB® are identical to those of the IKA 2000 inline production machines: UTL, DR, CMS, MHD, MK and MKO. Therefore, the magic LAB® ensures a reliable scale-up by offering the possibility to work with the same method from formulation development to mass production.
The DISPAX REACTOR (DR) module is a three-stage inline dispersing machine for the production of very fine emulsions and suspensions with a narrow particle spectrum. Thanks to the three rotor-stator combinations (generators) connected in series, it is suitable for both recirculation processes and fully continuous operation.
Optimum mixing , dispersing and wet milling results due to rotor tip speed up to 40 m/s and therefore high energy input into the working area. Easy and quick exchange of the modules as well as conversion to a complete working plant. Together with inline machines of 2000 series enables seamless transition of processes from laboratory to production while retaining the product quality.