ContactUsLinkComponent
Screw Closures for Narrow Neck Bottle Series 301 and 302, LDPE
Screw Closures for Narrow Neck Bottle Series 301 and 302, LDPE
Catalog # KAUT2000090603
Supplier:  Kautex
undefined
Screw Closures for Narrow Neck Bottle Series 301 and 302, LDPE
Catalog # KAUT2000090603
Supplier:  Kautex
Specifications
  • Thread:
    14 mm
  • Colour:
    Natural
  • Material:
    LDPE
  • Pk:
    1
  • For bottles:
    10/20/30 ml
Specifications
About this item
LDPE screw closures are packaged in polyethylene bags.