Graphite GC Ferrules
About this item
Graphite ferrules are commonly used in GC systems on the inlet and detector ends because they don’t stick to the column, and can be removed and reinserted easily. They should not be used with GC/MS instruments because of permeability to air.
- Made from high purity exfoliated graphite
- Seals with minimal torque and can be frequently reused if not over compressed
- Graphite ferrules do not shrink in use but are very soft and must be packed and handled with care
You should always trim a short piece of the column after replacing the ferrule to be sure that not graphite particles have plugged the column.
Upper temperature limit of 450 °C.
46 Options Available Below
Product Details & Documents
Graphite ferrules are commonly used in GC systems on the inlet and detector ends because they don’t stick to the column, and can be removed and reinserted easily. They should not be used with GC/MS instruments because of permeability to air.
- Made from high purity exfoliated graphite
- Seals with minimal torque and can be frequently reused if not over compressed
- Graphite ferrules do not shrink in use but are very soft and must be packed and handled with care
You should always trim a short piece of the column after replacing the ferrule to be sure that not graphite particles have plugged the column.
Upper temperature limit of 450 °C.
Product Family Options
Specialty GC ferrules
Product Information
- MaterialØ int.AvailabilityPrice
- SGEA211982-10Cup GC ferrule for Thermo Finnigan (M4 nut)Cup GC ferrule for Thermo Finnigan (M4 nut)Graphite0,28 mmLog in for availability1 * 10 items73,30 €
- SGEA211983-10Cup GC ferrule for Thermo Finnigan (M4 nut)Cup GC ferrule for Thermo Finnigan (M4 nut)Graphite0,38 mmLog in for availability1 * 10 items73,30 €
- SGEA211174Shimadzu GC ferrule, 17A, 2010, and 2014, two piece graphiteShimadzu GC ferrule, 17A, 2010, and 2014, two piece graphiteGraphite0,4 mmLog in for availability1 * 10 items93,30 €
- SGEA211984-10Cup GC ferrule for Thermo Finnigan (M4 nut)Cup GC ferrule for Thermo Finnigan (M4 nut)Graphite0,48 mmLog in for availability1 * 10 items73,30 €
- SGEA211175Shimadzu GC ferrule, 17A, 2010, and 2014, two piece graphiteShimadzu GC ferrule, 17A, 2010, and 2014, two piece graphiteGraphite0,5 mmLog in for availability1 * 10 items93,30 €
- SGEA211178Shimadzu GC ferrule, 17A, 2010, and 2014, two piece graphiteShimadzu GC ferrule, 17A, 2010, and 2014, two piece graphiteGraphite0,8 mmLog in for availability1 * 10 items93,30 €
- SGEA211988-10Cup GC ferrule for Thermo Finnigan (M4 nut)Cup GC ferrule for Thermo Finnigan (M4 nut)Graphite0,80 mmLog in for availability1 * 10 items73,30 €
- SGEA211500Shimadzu GC ferrule graphite
- SGEA211550Liner GC ferrule for Varian 1078/1079
Product Information
- MaterialØ int.AvailabilityPrice
- SGEA211103Long GC ferrule 1,58 mm
- SGEA211104Long GC ferrule 1,58 mm
- SGEA211164Agilent style GC ferrule 1,58 mm
- SGEA211164-200Agilent style GC ferrule 1,58 mm
- SGEA211204GC ferrule 3,17 mm
- SGEA211904GC ferrule 0,4 mm for ThermoFinnigan (M8 nut)GC ferrule 0,4 mm for ThermoFinnigan (M8 nut)Graphite0,4 mmLog in for availability1 * 10 items40,80 €
- SGEA211105Long GC ferrule 1,58 mm
- SGEA211125GC ferrule 1,58 mm, two hole graphite
- SGEA211165Agilent style GC ferrule 1,58 mm
- SGEA211205GC ferrule 3,17 mm
- SGEA211405GC ferrule 6,5 mm
- SGEA211905GC ferrule 0,5 mm for ThermoFinnigan (M8 nut)GC ferrule 0,5 mm for ThermoFinnigan (M8 nut)Graphite0,5 mmLog in for availability1 * 10 items40,80 €
- SGEA211108Long GC ferrule 1,58 mm
- SGEA211168Agilent style GC ferrule 1,58 mm
- SGEA211208GC ferrule 3,17 mm
- SGEA211408GC ferrule 6,35 mm
- SGEA211908GC ferrule 0,8 mm for ThermoFinnigan (M8 nut)GC ferrule 0,8 mm for ThermoFinnigan (M8 nut)Graphite0,8 mmLog in for availability1 * 10 items40,80 €
- SGEA211110Long GC ferrule 1,58 mm
- SGEA211160Agilent style GC ferrule 1,58 mm
- SGEA211201GC ferrule 3,17 mm
- SGEA211112Long GC ferrule 1,58 mm
- SGEA211100Long GC ferrule 1,58 mm
- SGEA211170Agilent style GC ferrule 1,58 mm
- SGEA211210GC ferrule 3,17 mm
- SGEA211410GC ferrule 6,35 mm
- SGEA211200GC ferrule 3,17 mm
- SGEA211440GC ferrule 6,35 mm
- SGEA211406NLGC ferrule 6,35 mm
- SGEA211406GC ferrule 6,5 mm
- SGEA211400GC ferrule 6,35 mm
- SGEA211600GC ferrule 9,525 mm
- SGEA211800GC ferrule 12,7 mm
- SGEA211120GC ferrule 19,05 mm
- SGEA211420GC ferrule 6,35 mm
- SGEA2111XXGC ferrule 1,58 mm, no hole
- SGEA2112XXGC ferrule 3,17 mm, no hole
- SGEA2114XXGC ferrule 6,5 mm, no hole