Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Weights
Calibration Weights
Calibration Weights, Class M1
Calibration Weights, Class M1
Catalog # ZWIEP2054512
Supplier:
ZWIEBEL
Expand image
Calibration Weights, Class M1
Catalog # ZWIEP2054512
Supplier:
ZWIEBEL
Specifications
Weight:
20 kg
Description:
Calibration weights, stainless steel, rectangular
Pk:
1
Tolerance:
±1000 mg
Specifications
About this item
Product Details
Product Details