Maintenaince Kit for Star Desiccators
Catalog # BOHLV1778-02
Supplier: Bohlender
Specifications
- Description:Maintenaince kit
- Pk:1 KIT
- For:SICCO Star desiccators
Specifications
About this item
Maintenance Kit for SICCO Star-Desiccators consists of one cellular rubber sealing tape, one magnetic tape and four non-slip rubber feet.
- Easy replacement of sealing and magnetic tape to ensure the airtight closure of the desiccator