Orders LinkContactUsLinkComponent
Spoon, Open
Spoon, Open
200 mm
Catalog # USBE3408
Supplier:   USBECK Laborgeräte
Spoon, Open
Catalog # USBE3408
Spoon, Open
Spoon, Open
200 mm
Price
4,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Spoon with open end made from stainless steel.
  • Autoclavable
  • Easy to clean
  • 25 mm width
  • 29 g weight
Excellent chemical resistance. High temperature resistance.
Specifications
  • Length:
    200 mm