HIGH STAT PLUS ESD Labcoats
Unisex ESD labcoat with long sleeves.
- Reverse collar
- Centre front closure with hidden press studs
- 1 chest pocket on the left
- 1 inside pocket at chest level on the right
- 2 patched side pockets
- Inset sleeves, adjustable with press studs at the bottom
- ESD logo
Industrial launderable.
: Oeko-Tex® standard 100; IEC 61340 - 4 - 9:2016.
- Catalog No:
- ALSO1000547-XL
- ALSO1002183-XS
- ALSO1000547-2XL
- ALSO1000547-L
- ALSO1000709-2XL
- ALSO1002183-2XL
- ALSO1000709-XS
- ALSW1031122-S
- ALSO1000547-XS
- ALSO1000547-S
- ALSW1031122-M
- ALSO1000547-M
- ALSO1002183-S
- ALSO1002183-M
- ALSO1002183-L
- ALSO1002183-XL
- Garment application:
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Cleanroom, Industrial
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Fabric:
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- 68% polyester/30,5% cotton/1,5% carbon
- Weight:
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
- 175 g/m²
Frequently Bought Together