ContactUsLinkComponent
Scotch® 233 Masking Tape
Scotch® 233 Masking Tape
Catalog # MMMA6355
Supplier:  3M
undefined
Scotch® 233 Masking Tape
Catalog # MMMA6355
Supplier:  3M
Specifications
  • Width:
    24 mm
  • Colour:
    Beige
  • Description:
    Scotch® Masking tape, 233
  • Length:
    50 m
  • Pk:
    36 Roll
Specifications
About this item
Scotch® Masking tape 233 is a premium masking tape, made of very thin beige crepe paper. This high-performance masking tape has excellent solvent and water resistance properties and is particularly suited to high temperature exposure.
  • High temperature resistance; up to 120 °C for up to 1 hour
  • Excellent solvent resistance
  • Conformable tape hugs curves and contours
  • Easy to apply and fast and clean to remove
  • Thin, tough beige crepe paper base
The high-performance Scotch® Masking Tape 233 is designed for automotive paint refinish applications. It is conformable, hugs curves and contours, applies quickly and easily, and removes cleanly. It has a particular high temperature resistance and is water- and solvent-resistant.