ContactUsLinkComponent
Cleanroom Earloop Facemasks with Clips, Non Sterile, Peta 230
Cleanroom Earloop Facemasks with Clips, Non Sterile, Peta 230
Catalog # 114-9595
Supplier:  ISOFIELD IRELAND
undefined
Cleanroom Earloop Facemasks with Clips, Non Sterile, Peta 230
Catalog # 114-9595
Supplier:  ISOFIELD IRELAND
Specifications
  • Type:
    With earloops and clips
  • Colour:
    White
  • Size:
    Universal
  • Environmentally Preferable:
  • Pk:
    600
Specifications
About this item
The Isofield Peta facemask is ISO Class 4 compatible.
  • ISO Class 4 compatible
  • Extra wide at 230 mm
  • Earloops with clip
  • Individually packed
  • Low CFU
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable ear loops, the Peta is also fitted with a convenient clip. The individually packed Peta facemasks provide excellent bacterial and particle filtration efficiency.
Packaging: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag, 12 inner bags per case (600 pieces).