ContactUsLinkComponent
Cleanroom Earloop Facemasks with Clips, Sterile, Exa 230
Cleanroom Earloop Facemasks with Clips, Sterile, Exa 230
Catalog # 114-9593
Supplier:  ISOFIELD IRELAND
undefined
Cleanroom Earloop Facemasks with Clips, Sterile, Exa 230
Catalog # 114-9593
Supplier:  ISOFIELD IRELAND
Specifications
  • Type:
    With earloops and clips
  • Colour:
    White
  • Size:
    Universal
  • Environmentally Preferable:
  • Pk:
    600
Specifications
About this item
The Isofield Exa is the widest validated sterile facemask available.
  • ISO Class 4, GMP Grade A compatible
  • Extra wide at 230 mm
  • Earloops with clip
  • Validated sterile
  • No-smell gamma compatible materials
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable ear loops, the Exa is also fitted with a convenient clip to adapt to your SOP, and is made with gamma compatible materials to ensure low/no odour in use. These individually packed Exa facemasks provide excellent bacterial and particle filtration efficiency, and are GMP Grade A compatible.
Packaging: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag, 12 inner bags per case (600 pieces).