Orders LinkContactUsLinkComponent
Baskets and Accessories for Modular Basket Systems
Supplier:   ELMA SCHMIDBAUER
Baskets and Accessories for Modular Basket Systems
Supplier:  ELMA SCHMIDBAUER
Certificates
Baskets and Accessories for Modular Basket Systems
30 Options Available Below
Product Details & Documents
These accessories for the modular basket system for Elmasonic P, Elmasonic select, EASY make cleaning small parts easy. These components for use with the Modular Basket System allow users to combine, adapt and customise the basket system for their requirements for cleaning with for Elmasonic P, Elmasonic Select and EASY ultrasonic baths.
  • Basket for cleaning small parts
  • Compatible with Elma pins
  • Modular Basket System is a powerful tool enabling effective and efficient cleaning processes as well as flexible loading
  • The number of objects that can be cleaned in the basket is maximised
  • Optimal cleaning results are ensured thanks to perfect placement
  • Helps save water, cleaning chemicals, energy as well as time and money
Specifications for Product Family
Specifications
  • Catalog No:
  • ELMA3000006016
  • ELMA3000006018
  • ELMA1125224
  • ELMA1125222
  • ELMA1125223
  • ELMA1125221
  • ELMA1125220
  • ELMA1125215
  • ELMA1125216
  • ELMA1125210
  • ELMA1125209
  • ELMA1125212
  • ELMA1125214
  • ELMA1125211
  • ELMA1125213
  • ELMA1124042
  • ELMA1125225
  • ELMA1125227
  • ELMA1125226
  • ELMA1125228
  • ELMA3000006017
  • ELMA3000006013
  • ELMA3000006014
  • ELMA3000006015
  • ELMA3000009104
  • ELMA3000006012
  • ELMA1125217
  • ELMA1125218
  • ELMA1125208
  • ELMA1125219
  • Basket load:
  • 2
  • 1
  • 2
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • ELMA3000006016
    Cover for minature round basket, stainless steel, Ext.Ø×H: 20×4 mm
    Cover for minature round basket, stainless steel, Ext.Ø×H: 20×4 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 18,00 €
    18,00 €
    1 * 1 items
       18,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006016
    Basket load
  • ELMA3000006018
    Cover for round basket, stainless steel, Ext.Ø×H: 64×2 mm
    Cover for round basket, stainless steel, Ext.Ø×H: 64×2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 14,00 €
    14,00 €
    1 * 1 items
       14,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006018
    Basket load
  • ELMA1125224
    Dividing wall set L high. 2 sets for basket sizes: 180/300, hole Ø 10×10 mm, stainless steel, 255×130×1 mm
    Dividing wall set L high. 2 sets for basket sizes: 180/300, hole Ø 10×10 mm, stainless steel, 255×130×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    1 * 1 items
       41,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125224
    Basket load
  • ELMA1125222
    Dividing wall set L low. 4 sets for basket sizes: 150, hole Ø 10×10 mm, stainless steel, 200×90×1 mm
    Dividing wall set L low. 4 sets for basket sizes: 150, hole Ø 10×10 mm, stainless steel, 200×90×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 36,00 €
    36,00 €
    1 * 1 items
       36,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125222
    Basket load
  • ELMA1125223
    Dividing wall set M. 2 sets for basket sizes: 100/120, hole Ø 10×10 mm, stainless steel, 200×60×1 mm
    Dividing wall set M. 2 sets for basket sizes: 100/120, hole Ø 10×10 mm, stainless steel, 200×60×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 36,00 €
    36,00 €
    1 * 1 items
       36,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125223
    Basket load
  • ELMA1125221
    Dividing wall set S high. 2 sets for basket sizes: 40/60/100/120/180 and 4 sets for basket sizes: 80/300, hole Ø 10×10 mm, stainless steel, 105×90×1 mm
    Dividing wall set S high. 2 sets for basket sizes: 40/60/100/120/180 and 4 sets for basket sizes: 80/300, hole Ø 10×10 mm, stainless steel, 105×90×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    1 * 1 items
       41,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125221
    Basket load
  • ELMA1125220
    Dividing wall set S low. 1 set for basket size: 20; 2 sets for basket sizes: 30/40/60/100/120/180 and 4 sets for basket sizes: 80/150/300, hole Ø 10×10 mm, stainless steel, 105×50×1 mm
    Dividing wall set S low. 1 set for basket size: 20; 2 sets for basket sizes: 30/40/60/100/120/180 and 4 sets for basket sizes: 80/150/300, hole Ø 10×10 mm, stainless steel, 105×50×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 26,00 €
    26,00 €
    1 * 1 items
       26,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125220
    Basket load
  • ELMA1125215
    Elma Pin 10×0001R, plastic, Ext.Ø×H: 40×75 mm, Int.Ø×H: 30×65 mm
    Elma Pin 10×0001R, plastic, Ext.Ø×H: 40×75 mm, Int.Ø×H: 30×65 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125215
    Basket load
  • ELMA1125216
    Elma Pin 10×0021R, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 15×40 mm
    Elma Pin 10×0021R, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 15×40 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125216
    Basket load
  • ELMA1125210
    Elma Pin 10×0158, plastic, Ext.Ø×H: 50×40 mm, Int.Ø×H: 40×25 mm
    Elma Pin 10×0158, plastic, Ext.Ø×H: 50×40 mm, Int.Ø×H: 40×25 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125210
    Basket load
  • ELMA1125209
    Elma Pin 10×0509, black, plastic, Ext.Ø×H: 30×35 mm, Int.Ø×H: 20×20 mm
    Elma Pin 10×0509, black, plastic, Ext.Ø×H: 30×35 mm, Int.Ø×H: 20×20 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125209
    Basket load
  • ELMA1125212
    Elma Pin 10×0613 and 0614, plastic, Ext.Ø×H: 55×45 mm and 75×45 mm, Int.Ø×H: 0,35×25 mm and 25×25 mm
    Elma Pin 10×0613 and 0614, plastic, Ext.Ø×H: 55×45 mm and 75×45 mm, Int.Ø×H: 0,35×25 mm and 25×25 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 51,00 €
    51,00 €
    1 * 1 items
       51,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125212
    Basket load
  • ELMA1125214
    Elma Pin 10×0741, plastic, Ext.Ø×H: 25×30 mm, Int.Ø×H: 4×15 mm
    Elma Pin 10×0741, plastic, Ext.Ø×H: 25×30 mm, Int.Ø×H: 4×15 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125214
    Basket load
  • ELMA1125211
    Elma Pin 10×0767, plastic, Ext.Ø×H: 85×65 mm, Int.Ø×H: 50×35 mm
    Elma Pin 10×0767, plastic, Ext.Ø×H: 85×65 mm, Int.Ø×H: 50×35 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125211
    Basket load
  • ELMA1125213
    Elma Pin 10×0772, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 5×40 mm
    Elma Pin 10×0772, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 5×40 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125213
    Basket load
  • ELMA1124042
    Elma starter set (contains: 3× each Elma pin 0741 and 0772 per set, 2× each Elma Pin 0509, 0158, 0767, 0613 and 0614, 0021R and 0001R per set, 2× silicone holder side comb and bottom comb each per set)
    Elma starter set (contains: 3× each Elma pin 0741 and 0772 per set, 2× each Elma Pin 0509, 0158, 0767, 0613 and 0614, 0021R and 0001R per set, 2× silicone holder side comb and bottom comb each per set)
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 51,00 €
    51,00 €
    1 * 1 items
       51,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1124042
    Basket load
  • ELMA1125225
    Level set type S with one basket load, hole Ø 11×15 mm, stainless steel, 195×105×1 mm
    Level set type S with one basket load, hole Ø 11×15 mm, stainless steel, 195×105×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 18,00 €
    18,00 €
    1 * 1 items
       18,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125225
    Basket load
  • ELMA1125227
    Level set type L, hole Ø 11×15 mm, stainless steel, 250×195×1 mm
    Level set type L, hole Ø 11×15 mm, stainless steel, 250×195×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 33,00 €
    33,00 €
    1 * 1 items
       33,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125227
    Basket load
    2
  • ELMA1125226
    Level set type M, hole Ø 11×15 mm, stainless steel, 255×105×1 mm
    Level set type M, hole Ø 11×15 mm, stainless steel, 255×105×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125226
    Basket load
    1
  • ELMA1125228
    Level set type XL, hole Ø 11×15 mm, stainless steel, 285×250×1 mm
    Level set type XL, hole Ø 11×15 mm, stainless steel, 285×250×1 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 36,00 €
    36,00 €
    1 * 1 items
       36,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125228
    Basket load
    2
  • ELMA3000006017
    Minature round basket without divisions, stainless steel, Ext.Ø×H: 21×12 mm, Int.Ø×H: 18×11 mm
    Minature round basket without divisions, stainless steel, Ext.Ø×H: 21×12 mm, Int.Ø×H: 18×11 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 14,00 €
    14,00 €
    1 * 1 items
       14,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006017
    Basket load
  • ELMA3000006013
    Round basket with divisions creating 4 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Round basket with divisions creating 4 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 36,00 €
    36,00 €
    1 * 1 items
       36,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006013
    Basket load
  • ELMA3000006014
    Round basket with divisions creating 5 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Round basket with divisions creating 5 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    1 * 1 items
       41,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006014
    Basket load
  • ELMA3000006015
    Round basket with divisions creating 9 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Round basket with divisions creating 9 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 64,00 €
    64,00 €
    1 * 1 items
       64,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006015
    Basket load
  • ELMA3000009104
    Round basket without divisions, stainless steel, ¹/₁ height, Ext.Ø×H: 64×42 mm, Int.Ø×H: 60×41 mm
    Round basket without divisions, stainless steel, ¹/₁ height, Ext.Ø×H: 64×42 mm, Int.Ø×H: 60×41 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    1 * 1 items
       41,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000009104
    Basket load
  • ELMA3000006012
    Round basket without divisions, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Round basket without divisions, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 22,00 €
    22,00 €
    1 * 1 items
       22,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA3000006012
    Basket load
  • ELMA1125217
    Silicone holder 10×bottom comb set, 105×10×35 mm
    Silicone holder 10×bottom comb set, 105×10×35 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 62,00 €
    62,00 €
    1 * 1 items
       62,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125217
    Basket load
  • ELMA1125218
    Silicone holder 10×side comb set, 150×10×45 mm
    Silicone holder 10×side comb set, 150×10×45 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 62,00 €
    62,00 €
    1 * 1 items
       62,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125218
    Basket load
  • ELMA1125208
    Silicone holder set (10), Ext.Ø×H: 20×20 mm
    Silicone holder set (10), Ext.Ø×H: 20×20 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 16,00 €
    16,00 €
    1 * 1 items
       16,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125208
    Basket load
  • ELMA1125219
    Holder set, stainless steel
    Holder set, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,00 €
    41,00 €
    1 * 1 items
       41,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    ELMA1125219
    Basket load