ContactUsLinkComponent
Charging base for Rainin AutoRep™ E
Catalog # RAINAR-RB
Supplier:  RAININ
Charging base for Rainin AutoRep™ E
Catalog # RAINAR-RB
Supplier:  RAININ
Specifications
  • Description:
    Charging base for battery, for AutoRep™ E
  • Pk:
    1
Specifications
About this item
Recharging base for AutoRep™ E battery pack. Accepts complete AutoRep E with battery installed (quicker charge), or battery alone (slower charge).