ContactUsLinkComponent
Carousel stand for 7 pipettes, Rainin CR-7
Carousel stand for 7 pipettes, Rainin CR-7
Catalog # 613-4662
Supplier:  RAININ
Carousel stand for 7 pipettes, Rainin CR-7
Catalog # 613-4662
Supplier:  RAININ
Specifications
  • Description:
    Carousel stand, white, for 7 Rainin™ manual pipettes and Gilson® Pipetman® / Rainin electronic and multi-channel pipettes (adapter 17006638 needed)
  • Pk:
    1
Specifications
About this item
Carousel stand, holds up to 7 Rainin™ manual pipettes and Gilson® Pipetman®, as well as Rainin electronic and multi-channel pipettes with adapter 17006638 (HU-A3).
  • Keep your pipettes ready for use, upright and safe
  • Weighted base keeps the stand upright and prevents tipping
  • Needs little bench space, rotate and pull the pipette you need