Order Entry
Slovenia
ContactUsLinkComponent
Anti-BCL2L15 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-BCL2L15 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7582R-A750
Supplier:  Bioss
Anti-BCL2L15 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7582R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCL2L15
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    65-163/163
  • Form:
    Liquid
  • Gene ID:
    440603
  • Antigen synonyms:
    Bcl2-L-15|pro-apoptotic Bcl-2 protein|Bfk|C1orf178|Bcl-2-like protein 15|B2L15_HUMAN|FLJ22588|Bcl2l15|BCL2-like 15
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    18 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human BCL2L15
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

BCL2L15.

Type: Primary
Antigen: BCL2L15
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 65-163/163
Host: Rabbit
Isotype: IgG
Reactivity: Human