Specifications
- Antibody type:Primary
- Antigen name:alpha-TAT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:1-100/421
- Form:Liquid
- Gene ID:79969
- Antigen synonyms:Alpha tubulin N acetyltransferase|Hypothetical protein LOC79969|Acetyltransferase mec 17 homolog|Alpha TAT|MEC17|Nbla00487|Alpha tubulin acetyltransferase 1|ATAT_HUMAN|TAT|Chromosome 6 open reading frame 134|C6orf134
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human alpha-TAT
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Making up nearly 6% of the human genome, chromosome 6 contains around 1200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatibility complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6. The C6orf134 gene product has been provisionally designated C6orf134 pending further characterisation.
Type: Primary
Antigen: alpha-TAT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/421
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat