Anti-Gamma-Adaptin Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1600R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Gamma-Adaptin
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:721-822/822
- Form:Liquid
- Gene ID:11765
- Antigen synonyms:Adaptor-related protein complex 1 subunit gamma-1|Ap1g1|Adaptor protein complex AP-1 subunit gamma-1|Golgi adaptor HA1/AP1 adaptin subunit gamma-1|Adtg|Gamma1-adaptin|AP-1 complex subunit gamma-1|Gamma-adaptin|Clapg1|Clathrin assembly protein complex 1 gamma-1 large chain
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:91 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse Gamma-Adaptin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Subunit of clathrin-associated adaptor protein complex 1 that plays a role in protein sorting in the late-Golgi/trans-Golgi network (TGN) and/or endosomes. The AP complexes mediate both the recruitment of clathrin to membranes and the recognition of sorting signals within the cytosolic tails of transmembrane cargo molecules.
Type: Primary
Antigen: Gamma-Adaptin
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 721-822/822
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat