Anti-RBBP6/P2P-R Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-1483R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RBBP6/P2P-R
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:2-150/1792
- Form:Liquid
- Gene ID:5930
- Antigen synonyms:PACT|SNAMA|E3 ubiquitin-protein ligase RBBP6|Retinoblastoma-binding protein 6|RBBP6|Protein P2P-R|MY038|RBQ1|p53-associated cellular protein of testis|RBQ-1|P2P-R|Retinoblastoma-binding Q protein 1|Proliferation potential-related protein|P2PR
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:201 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human P2PR
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
E3 ubiquitin-protein ligase which promotes ubiquitination of YBX1, leading to its degradation by the proteasome. May play a role as a scaffold protein to promote the assembly of the p53/TP53-MDM2 complex, resulting in increase of MDM2-mediated ubiquitination and degradation of p53/TP53; may function as negative regulator of p53/TP53, leading to both apoptosis and cell growth.
Type: Primary
Antigen: RBBP6/P2P-R
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 2-150/1792
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat