Order Entry
Slovenia
ContactUsLinkComponent
Anti-RBBP6/P2P-R Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-RBBP6/P2P-R Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-1483R-A488
Supplier:  Bioss
Anti-RBBP6/P2P-R Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-1483R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RBBP6/P2P-R
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    2-150/1792
  • Form:
    Liquid
  • Gene ID:
    5930
  • Antigen synonyms:
    PACT|SNAMA|E3 ubiquitin-protein ligase RBBP6|Retinoblastoma-binding protein 6|RBBP6|Protein P2P-R|MY038|RBQ1|p53-associated cellular protein of testis|RBQ-1|P2P-R|Retinoblastoma-binding Q protein 1|Proliferation potential-related protein|P2PR
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    201 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human P2PR
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

E3 ubiquitin-protein ligase which promotes ubiquitination of YBX1, leading to its degradation by the proteasome. May play a role as a scaffold protein to promote the assembly of the p53/TP53-MDM2 complex, resulting in increase of MDM2-mediated ubiquitination and degradation of p53/TP53; may function as negative regulator of p53/TP53, leading to both apoptosis and cell growth.

Type: Primary
Antigen: RBBP6/P2P-R
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 2-150/1792
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat