Anti-BOb-1/POU2AF1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1418R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BOb-1/POU2AF1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:21-130/256
- Form:Liquid
- Gene ID:5450
- Antigen synonyms:BOB-1|POU2AF1|B-cell-specic coactivator OBF-1|OBF1_HUMAN.|OCA-B|B-cell oct-binding protein 1|OCA-B. OBF1|POU domain class 2-associating factor 1|OCT-binding factor 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:28 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BOb-1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. This receptor has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. Its ligands included a variety of aromatic hydrocarbons.
Type: Primary
Antigen: BOb-1/POU2AF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 21-130/256
Host: Rabbit
Isotype: IgG
Reactivity: Rat