Specifications
- Antibody type:Primary
- Antigen name:p70 S6 Kinase Beta 2/P70 Beta 2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:165-270/482
- Form:Liquid
- Gene ID:6199
- Antigen synonyms:p70 S6 kinase beta|RPS6KB2|p70 ribosomal S6 kinase beta|S6K-beta|70 kDa ribosomal protein S6 kinase 2|S6K beta 2|S6K2|p70 S6KB|p70S6K beta|SRK|Ribosomal protein S6 kinase beta 2|Serine/threonine protein kinase 14 beta|p70S6Kb|S6 kinase related kinase|p70 S6Kbeta|Ribosomal protein S6 kinase 70kDa, polypeptide 2|S6K beta|p70-S6KB|Ribosomal protein S6 kinase beta-2|STK14B|p70-beta|Serine/threonine-protein kinase 14 beta|S6 kinase-related kinase|p70 beta
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human p70 S6 Kinase Beta 2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.
Type: Primary
Antigen: p70 S6 Kinase Beta 2/P70 Beta 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 165-270/482
Host: Rabbit
Isotype: IgG
Reactivity: Rat