Cleanroom gloves, polyisoprene, sterile, Biogel® PI Micro Tech Indicator® underglove
Supplier: MOLNLYCKE HEALTH CARE
Certificates
About this item
These sterile, blue cleanroom undergloves with beaded cuffs and curved fingers are made to be worn in combination with the Biogel® PI Micro Tech overglove to form a Breach Indication System. The gloves are suitable for use in aseptic and Class 100 (ISO 5)/EU GMP Grade A environments.
- Tested and cleared for use with chemotherapy agents
- Synthetic polyisoprene
- Exceed industry standard for force at break, tensile strength and elongation
- Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Ideal for use in controlled environments, in medical and research laboratories, in the pharma industry, as well as in biotechnology.
Certifications: EN 455, ASTM D3577, ISO 10282, EN 374, EN 16523-1, PPE Cat. III, AQL 0,65, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, ASTM F1671, Allergenicity ISO 10993, Pyrogenicity ASTM D7102, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420.
Packaging: One pair per pack, in a high quality polyethylene inner wrap, packed into a film pack. 25 pairs per inner LDPE polybag f; 2 inner LDPE polybags are packed in an outer LDPE polybag. Four outer polybags per transit case, total of 200 pairs.
Specifications
- Catalog No:
- 112-6323
- 112-6345
- 112-6324
- 112-6322
- 112-6325
- 112-6326
- 112-6327
- 112-6328
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- Finger thickness:
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- 0,21 mm
- Palm thickness:
- 0,19 mm
- 0,19 mm
- 0,19 mm
- 0,19 mm
- 0,19 mm
- 0,19 mm
- 0,19 mm
- 0,19 mm
- Tensile strength:
- 24 MPa
- 24 MPa
- 24 MPa
- 24 MPa
- 24 MPa
- 24 MPa
- 24 MPa
- 24 MPa
- Ultimate elongation:
- 1150
- 1150
- 1150
- 1150
- 1150
- 1150
- 1150
- 1150
- EN norm:
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 10282, EN ISO 374-1, EN 374-2, EN 374-4, EN 16523-1, EN ISO 374-5, EN 556-1, EN ISO 15223-1, EN ISO 21420
- Environmental certification:
- FCS, ISO 14001
- FCS, ISO 14001
- FCS, ISO 14001
- FCS, ISO 14001
- FCS, ISO 14001
- FCS, ISO 14001
- FCS, ISO 14001
- FCS, ISO 14001