ContactUsLinkComponent
Thread adapter AISI 304, G 1½" inner - G 2" inner
Thread adapter AISI 304, G 1½" inner - G 2" inner
Catalog # BURK0600-3100
Supplier:  BURKLE
undefined
Thread adapter AISI 304, G 1½" inner - G 2" inner
Catalog # BURK0600-3100
Supplier:  BURKLE
Specifications
  • Thread:
    G 1½" inner - G 2" inner
  • Material:
    Stainless steel
  • Pk:
    1
Specifications
About this item
With the thread adapter made of high-quality stainless steel 1.4301 (304) a pump, a drain cock, or any other connection with thread G 2" (Whitworth pipe thread) can be screwed onto a container with G 1½" thread.
  • Adapts male thread G 1½" to male thread G 2"
  • Made of high quality stainless steel
  • Suitable for flammable liquids
The thread adapter adapts and thus compensates for different thread shapes. The drum adapter ensures that decanting is liquid-tight; a seal made of ePTFE reliably seals the connection. The thread adapter made of stainless steel is electrically conductive and highly chemically resistant. It can therefore be used for filling solvents and other flammable liquids (e.g. alcohols, petrol, diesel etc.) as well as for weak alkalis and acids. Thread adapter for securely fastening Bürkle barrel screw connections, Bürkle barrel pumps, taps etc. to canisters, steel barrels, drums or Bürkle safety containers. The thread adapter can be used to screw the Bürkle solvent pumps onto the Bürkle stainless steel safety canisters and safety containers.