Rigid Conductive Boxes
About this item
Desco Europe Rigid Conductive Boxes are marked with the ESD protective symbol. Boxes can be supplied with or without high density foam for insertion of component leads or low density foam which acts as a cushioning material.
- Provide good ESD and mechanical protection
- Made of conductive polypropylene; Rs: 1×10² to <1×10⁵ Ω per IEC 61340-2-3
- Strong box moulded in one-piece
- Supplied with/without foam lining
- Top foam: 6 mm thick low density foam (FX)
- Bottom foam: 6 mm thick high density foam (HD)
Certifications: Meets EN 61340-5-1 and Packaging standard IEC 61340-5-3.
3 Options Available Below
Product Details & Documents
Desco Europe Rigid Conductive Boxes are marked with the ESD protective symbol. Boxes can be supplied with or without high density foam for insertion of component leads or low density foam which acts as a cushioning material.
- Provide good ESD and mechanical protection
- Made of conductive polypropylene; Rs: 1×10² to <1×10⁵ Ω per IEC 61340-2-3
- Strong box moulded in one-piece
- Supplied with/without foam lining
- Top foam: 6 mm thick low density foam (FX)
- Bottom foam: 6 mm thick high density foam (HD)
Certifications: Meets EN 61340-5-1 and Packaging standard IEC 61340-5-3.
Product Family Options
With foam lining
Product Information
- AvailabilityPrice
- 129-2620Rigid conductive box with foam in lid and base, 75×51×14 mmRigid conductive box with foam in lid and base, 75×51×14 mmLog in for availability1 * 1 items4,10 €
- 129-2619Rigid conductive box with foam in lid and base, 90×64×16 mmRigid conductive box with foam in lid and base, 90×64×16 mmLog in for availability1 * 1 items5,00 €
Without foam lining