Performance box sealing tape 3444, Scotch®
MMMA34445050
: 3M
- Width:50 mm
- Colour:Amber
- Length:50 m
- Environmentally Preferable:
- Model:3444
- Pk:36 Roll
Seal cartons by using Scotch® Performance Box Sealing Tape 3444. Our sealing tape is made from non-silicone flat back impregnated paper, coated with a natural rubber adhesive. It sticks to a wide variety of surfaces.
- Apply to carton sealing applications
- Use as a single strip closure of lightweight boxes, repaired and miscellaneous packaging
- Get non-silicone flat-back impregnated paper, coated with a high tack, natural rubber adhesive
- Good alternative to plastic tapes
- Good performance in damp and cold shipping and storage environments