96-Well high content screening microplates with film bottom, Corning®
Catalog # 734-2763
Supplier: Corning
Specifications
- No. of wells:96
- Sterile:+
- Colour:Black/clear
- Description:Half area microplate, with lid, tissue culture treated
- Seq:01
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Pk:16
- Well volume:200 µl
Specifications
About this item
With an ultraclear film bottom and excellent flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications.
- TC-treated, flat well bottom black microplate
- Made of cyclic olefin copolymer
- Ultraclear 190 µm film bottom thickness for imaging microscopy applications
- Low background fluorescence and minimal crosstalk allows highest possible optical quality for cell based assays
- Half area 96-well microplate reduces reagent consumption
- Sterile
- Inter and inter-well film bottom flatness optimised for high content applications
The microplate and film are manufactured from cyclic olefin copolymer, which has excellent optical properties, chemical resistance, and mechanical stability.