ContactUsLinkComponent
Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 2500
Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 2500
Catalog # ANSEWH25T-00705-05
Supplier:  Ansell
undefined
Encapsulated chemical protective suits for PAPR systems, AlphaTec® MICROGARD 2500
Catalog # ANSEWH25T-00705-05
Supplier:  Ansell
Specifications
  • Clothing Size:
    XL
  • Colour:
    White
  • Seq:
    04
  • Pk:
    5
Specifications
About this item
Encapsulated chemical protective suits for use with PAPR systems.
  • Provide complete protection from liquid and particulate hazards
  • Unique material offering exceptional mechanical strength, as well as liquid and particulate protection
  • Air permeable SMS collar maintains sufficient air in the breathing zone while allowing excess air to flow into the suit body
  • Four exhalation valves allow a full range of movement without risk of excessive pressure
  • Anti-static tested according to EN 1149-5
  • Designed for use with Malina CleanAir fan units (sold separately)
Ventilated suits with filtered air and MICROGARD technology provide head and body protection from hazardous substances. The MICROGARD PAPR range of suits are designed for use with Powered Air Purifying Respirators (PAPRs) and are certified to provide both respiratory and body protection from hazardous substances. Stitched and taped seams offer increased strength and an effective liquid and particle barrier.
Suits feature a front entry, panoramic visor design, double elasticated cuffs, air permeable SMS collar, and emergency rip cord.