Orders LinkContactUsLinkComponent
96-well plates, polypropylene, without lids
96-well plates, polypropylene, without lids
   Sustainable Options Available
Supplier:   Corning
96-well plates, polypropylene, without lids
Supplier:  Corning
   Sustainable Options Available
Certificates
96-well plates, polypropylene, without lids
9 Options Available Below
Product Details & Documents
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-1545
  • 734-1550
  • 734-1781
  • 734-1782
  • 734-1547
  • 734-1552
  • 734-1551
  • 732-0484
  • 732-0485
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
  • Well volume
    Sterile
    Packed
    Availability
    Price
  • 734-1545
    Storage plate, 96-well, natural, V-shaped well
    Storage plate, 96-well, natural, V-shaped well
    320 µl
    +
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 404,00 €
    404,00 €
    1 * 100 items
       404,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    734-1545
    Environmentally Preferable
    Environmental certification
  • 734-1550
    Storage plate, 96-well, natural, V-shaped well
    Storage plate, 96-well, natural, V-shaped well
    320 µl
    -
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 350,00 €
    350,00 €
    1 * 100 items
       350,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    734-1550
    Environmentally Preferable
    Environmental certification
  • 734-1781
    Storage plate, 96-well, white, round well
    Storage plate, 96-well, white, round well
    360 µl
    -
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 412,00 €
    412,00 €
    1 * 100 items
       412,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1781
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • 734-1782
    Storage plate, 96-well, black, round well
    Storage plate, 96-well, black, round well
    360 µl
    -
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 542,00 €
    542,00 €
    1 * 100 items
       542,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1782
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
      Low Manufacturing Impact
  • 734-1547
    Storage plate, 96-well, natural, round well
    Storage plate, 96-well, natural, round well
    360 µl
    +
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 398,00 €
    398,00 €
    1 * 100 items
       398,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    734-1547
    Environmentally Preferable
    Environmental certification
  • 734-1552
    Storage plate, 96-well, natural, round well
    Storage plate, 96-well, natural, round well
    360 µl
    -
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 350,00 €
    350,00 €
    1 * 100 items
       350,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    734-1552
    Environmentally Preferable
    Environmental certification
  • 734-1551
    Storage plate, 96-well, natural, flat well
    Storage plate, 96-well, natural, flat well
    360 µl
    -
    25/bag
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 350,00 €
    350,00 €
    1 * 100 items
       350,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    734-1551
    Environmentally Preferable
    Environmental certification
  • 732-0484
    Storage plate, 96-well, expanded volume, natural, V-bottom well
    Storage plate, 96-well, expanded volume, natural, V-bottom well
    450 µl
    -
    10/bag
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 327,00 €
    327,00 €
    1 * 50 items
       327,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-0484
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 732-0485
    Storage plate, 96-well, expanded volume, natural, V-bottom well
    Storage plate, 96-well, expanded volume, natural, V-bottom well
    450 µl
    +
    10/bag
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 373,00 €
    373,00 €
    1 * 50 items
       373,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-0485
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact