Activated carbon filter paper, Whatman™
Catalog # WHAT10320163
Supplier: Whatman products (Cytiva)
Specifications
- Pk:50
Specifications
About this item
Paper filter containing activated carbon, very helpful in retaining radioactive compounds from liquids or for the adsorption of iodine-131 from the air. Due to the adsorption properties of activated carbon, the paper can be used for each kind of clarification and brightening turbid media.