Specifications
- Sterile:+
- Garment application:
- Gender:
- Colour:White
- Environmentally Preferable:
- Pk:100
Specifications
About this item
These hoods with integrated mask, model IC689BWH WS, are made of Tyvek®, which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.
- Aseptically folded
- Ties for adjustable fit in the chest area
- Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
- Suitable for use in GMP class A/B (ISO Class 5) cleanrooms
The white hood features bound, internal seams and a face opening. Integrated mask is blue pleated polyethylene, measuring 175 mm wide. Note, mask is not a PPE garment (not FFP1, 2, or 3 mask).
All DuPont™ Tyvek® IsoClean® sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy-tear cleanroom bag.
Garments and accessories made of sterile Tyvek® IsoClean® are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments.