Order Entry
Slovenia
ContactUsLinkComponent
Oxalyl dichloride ≥97.5% for synthesis
Oxalyl dichloride ≥97.5% for synthesis
Catalog # 91303.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxalyl dichloride ≥97.5% for synthesis
Catalog # 91303.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    Min 97.5%

Specification Test Results

About this item

 :
 :126.93 g/mol
Boiling Pt:63…64 °C
Melting Pt:–12 °C
Density:1.478 g/cm³ (25 °C)
Storage Temperature:Refrigerator
EINECS:201-200-2
UN:3110
ADR:6.1+8,II
 :GHS05
 :GHS06
CAS Number:79-37-8
Formula:C₂Cl₂O₂
MDL Number:MFCD00000704