I-300 compatibility kit V-7X0
Catalog # BUCH11I300S40
Supplier: BUCHI
Specifications
- Description:Compatibility kit V-7X0 with 4,3" display
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Interface I-300 for use with BUCHI Rotavapour® R-21x/V-7x0 models.
- Control all process parameters from single controller
- Central control of all process parameters such as rotation speed, temperature (heating, cooling, vapour) and pressure
- Remote monitoring on smart phones including push notifications and live view
The Interface I-300 controls all distillation parameters of a Rotavapour system. Modes: manual, timer, cont., AutoDest, drying. Mobile app is available.
Features are: Interface: I-300 Compatibility Pack: R-210/R-215, V-700/V-710.