Lonicera Macranthoides Flower Dry Extract, Reference Standard
Catalog # 1369703.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:500 mg
- Standards:USP reference standard
- Description:Lonicera Macranthoides Flower Dry Extract
Specifications
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Mixture, with identified compendial components Dipsacoside B, Macranthoidin A and Macranthoidin B
Certifications: US Pharmacopea Reference Standard