Orders LinkContactUsLinkComponent
Riociguat, Reference Standards
Riociguat, Reference Standards
Supplier:   USP
Riociguat, Reference Standards
Supplier:  USP
Certificates
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Riociguat System Suitability Mixture: This is a mixture containing (approximately 0.2% of related compound B and 0.5% of related compound C) riociguat, riociguat related compound B and riociguat related compound C
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1604494.
  • 1604519.
  • 1604527.
  • Description:
  • Riociguat System Suitability Mixture II
  • Riociguat
  • Riociguat System Suitability Mixture
  • CAS No.:
  • 625115-55-1
Specifications
Product Family Options
Product Information
  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1604494.
    Riociguat System Suitability Mixture II
    Riociguat System Suitability Mixture II
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 240,00 €
    1 240,00 €
    1 * 25 mg
       1 240,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1604494.
    Description
    Riociguat System Suitability Mixture II
    CAS No.
  • 1604519.
    Riociguat
    Riociguat
    USP reference standard
    Vial
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 627,00 €
    627,00 €
    1 * 250 mg
       627,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1604519.
    Description
    Riociguat
    CAS No.
    625115-55-1
  • 1604527.
    Riociguat System Suitability Mixture
    Riociguat System Suitability Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 330,00 €
    1 330,00 €
    1 * 25 mg
       1 330,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1604527.
    Description
    Riociguat System Suitability Mixture
    CAS No.
Recommendations