Orders LinkContactUsLinkComponent
Dolasetron Mesylate, Reference Standards
 :   USP
Dolasetron Mesylate, Reference Standards
 :  USP
Dolasetron Mesylate, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
  • Catalog No:
  • 1224959.
  • 1224960.
  • Description:
  • Dolasetron Mesylate
  • Dolasetron Mesylate Related Compound A
  • Synonym(s):
  • Hexahydro-8-hydroxy-2,6-methano-2H-quinolizin-3 (4H)-one, hydrochloride
  • CAS No.:
  • 878143-33-0
  • 148091-18-3
Product Family Options
  • Description
    Standards
    Pack type
    Pk
  • Dolasetron Mesylate
    USP reference standard
    Vial
    125 mg
    1 * 125 mg
    1 * 125 mg - 295,00 €
    295,00 €
    1 * 125 mg
       295,00 €
    Specifications
    Cat. No.
    1224959.
    Description
    Dolasetron Mesylate
    Synonym(s)
    CAS No.
    878143-33-0
  • Dolasetron Mesylate Related Compound A
    USP reference standard
    Vial
    25 mg
    1 * 25 mg
    1 * 25 mg - 1 210,00 €
    1 210,00 €
    1 * 25 mg
       1 210,00 €
    Specifications
    Cat. No.
    1224960.
    Description
    Dolasetron Mesylate Related Compound A
    Synonym(s)
    Hexahydro-8-hydroxy-2,6-methano-2H-quinolizin-3 (4H)-one, hydrochloride
    CAS No.
    148091-18-3