Clipped corner microscope slide, Snowcoat®
Snowcoat® clipped corner slide, colored ends, White
Catalog # LEIC3800381
Supplier: LEICA MICROSYSTEMS
Jump to:
Product Details & Documents
Snowcoat® Clipped Corder Slides are clipped at 45° angles to help reduce glass breakage and fragmentation when loaded into slide printers.
- Available in a variety of colours
- Specialty painted surface
- Efficient scanning
Specialty painted surface formulated to optimise Ink Jet printing and bar code scanning efficiency. These slides have been optimised for use with the Leica printer.
Specifications
- Colour:White
- Description:Snowcoat® clipped corner slide, colored ends