Order Entry
Slovenia
ContactUsLinkComponent
Anti-VIP mouse monoclonal antibody
Anti-VIP mouse monoclonal antibody
Catalog # NOVUNBP1-05163
Supplier:  Novus Biologicals
Anti-VIP mouse monoclonal antibody
Catalog # NOVUNBP1-05163
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vasoactive Intestinal Peptide
  • Antigen symbol:
    VIP
  • Clonality:
    Monoclonal
  • Clone:
    2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Pig,
    Human,
    Bovine,
    Rat
  • Western blot:
    Yes
  • Gene ID:
    7432
  • Antigen synonyms:
    VIP peptides|vasoactive intestinal peptide|MGC13587|PHM27
  • Storage buffer:
    10 mM Phosphate (pH 7,4) and 0,5 M NaCl
  • Storage temperature:
    Store at 4 °C in the dark.
  • Concentration:
    1,0 mg/ml
  • Immunogen:
    Vasoactive intestinal peptide adsorbed onto aluminum hydroxide gel
  • Purification:
    Protein A or G purified
  • Pk:
    100 µl

Specifications

About this item

The VIP Antibody (2) - BSA Free from Novus Biologicals is a mouse monoclonal antibody to VIP. This antibody reacts with human, rat, porcine, bovine. The VIP Antibody (2) - BSA Free has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VIP
Clonality: Monoclonal
Clone: 2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Rat, Porcine, Bovine