Goat Polyclonal anti-Rabenosyn 5 antibody
Catalog # NOVUNB300-813
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Rabenosyn 5
- Antigen symbol:RBSN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:64145
- Antigen synonyms:Rabenosyn-5|FYVE-finger-containing Rab5 effector protein rabenosyn-5|FYVE domain containing 20|zinc finger|110 kDa protein|FYVE finger-containing Rab5 effector protein rabenosyn-5|FLJ34993|MGC126210|Zinc finger FYVE domain-containing protein 20
- Storage buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage temperature:Store at −20 °C. Avoid freeze-thaw cycles.
- Concentration:0,5 mg/ml
- Immunogen:Peptide with sequence C-ELKHTLAKQKGGTD corresponding to C-Terminus according to NP_071735.2.
- Tested applications:Peptide
- Purification:Immunogen affinity purified
- Pk:0,1 mg
Specifications
About this item
The Rabenosyn 5 Antibody from Novus Biologicals is a goat polyclonal antibody to Rabenosyn 5. This antibody reacts with human. The Rabenosyn 5 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.
Type: Primary
Antigen: Rabenosyn 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human