Anti-HIV1 p24 Goat polyclonal antibody
Catalog # NOVUNB100-65514
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:HIV-1 p24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Gene ID:155348
- Antigen synonyms:Capsid protein p24|Human immunodeficiency virus 1|Human immunodeficiency virus type 1 p24
- Storage buffer:No buffer
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:4,0 mg/ml
- Immunogen:Purified native p24, from strain IIIB.
- Purification:IgG purified
- Pk:0,5 mL
Specifications
About this item
Goat Polyclonal HIV-1 p24 Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Virus.
Type: Primary
Antigen: HIV1 p24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Virus