Masterflex® Printed Microscope Slide, ten rectangular sections; 144/pk
Catalog # MFLX75955-31
Supplier: Avantor Fluid Handling
Specifications
- Description:Microscope slides, printed with mask, ten rectangular sections
- L×W:76,2×25,4 mm
- Environmentally Preferable:
- Pk:144
- Slide type:Printed without Mask
Specifications
About this item
Optimize sample testing and viewing.
- Screen-printed section design keeps samples contained
- Hydrophobic epoxy ink withstands frequent washing and resists chemicals
- Contrasting ink makes orienting the microscope field of view over a sample easy
Printed microscope slides are available with a variety of section shapes, sizes, counts, colors, and labels. The PTFE-based epoxy ink creates a hydrophobic barrier, holding samples in place for easy viewing under a microscope, and keeping reactions isolated from each other in multi-section slides.
- Description:Microscope slides, printed with mask, ten rectangular sections
- L×W:76,2×25,4 mm
- Environmentally Preferable:
- Pk:144
- Slide type:Printed without Mask