Orders LinkContactUsLinkComponent
VWR®, Shell Vials
VWR®, Shell Vials
VWR®, Shell Vials
Supplier:  VWR Collection
Certificates
VWR®, Shell Vials
VWR®, Shell Vials
11 Options Available Below
Product Details & Documents
Shell vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a PE plug featuring a starburst top for easier needle penetration.
  • Economical choice for many HPLC applications
  • Used in combination with PE Plugs
  • Quick assembly
Shell vials constructed from type I borosilicate glass with low metal content. Vials are available in three different sizes (1, 2 and 4 ml) in amber or clear version with a flat bottom to ensure compatibility with inserts.
Product Family Options
Vials
Product Information
  • Capacity
    Base
    Ø×H
    Material
    Colour
    Cap included
    Availability
    Price
  • 548-0042A
    Shell vials, 8 mm PE plug with insertion barrier
    Shell vials, 8 mm PE plug with insertion barrier
    1,00 ml
    Flat bottom
    8×40 mm
    Borosilicate glass 5.1
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 367,00 €
    367,00 €
    1 * 1.000 items
       367,00 €
  • 548-0043A
    Shell vials, 8 mm PE plug with insertion barrier
    Shell vials, 8 mm PE plug with insertion barrier
    1,00 ml
    Flat bottom
    8×40 mm
    Borosilicate glass 5.1
    Amber
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 429,00 €
    429,00 €
    1 * 1.000 items
       429,00 €
  • 548-0347A
    Shell vials, 6 mm PE plug
    Shell vials, 6 mm PE plug
    1,00 ml
    Flat bottom
    7,8×35 mm
    Borosilicate glass 5.1
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 547,00 €
    547,00 €
    1 * 1.000 items
       547,00 €
  • 548-0352A
    Shell vials, 8 mm PE plug
    Shell vials, 8 mm PE plug
    1,00 ml
    Flat bottom
    8×40 mm
    Borosilicate glass 5.1
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 387,00 €
    387,00 €
    1 * 1.000 items
       387,00 €
  • 548-0353A
    Shell vials, 8 mm PE plug
    Shell vials, 8 mm PE plug
    1,00 ml
    Flat bottom
    8×40 mm
    Borosilicate glass 5.1
    Amber
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 429,00 €
    429,00 €
    1 * 1.000 items
       429,00 €
  • 548-2481A
    Shell vials without plug
    Shell vials without plug
    1,00 ml
    Flat bottom
    8×40 mm
    Borosilicate glass 5.1
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 350,00 €
    350,00 €
    1 * 1.000 items
       350,00 €
  • 548-0045A
    Shell vials, 12 mm PE plug
    Shell vials, 12 mm PE plug
    2,00 ml
    Flat bottom
    12×31,5 mm
    Borosilicate glass 5.1
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 546,00 €
    546,00 €
    1 * 1.000 items
       546,00 €
  • 548-0046A
    Shell vials, 12 mm PE plug
    Shell vials, 12 mm PE plug
    2,00 ml
    Flat bottom
    12×31,5 mm
    Borosilicate glass 5.1
    Amber
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 634,00 €
    634,00 €
    1 * 1.000 items
       634,00 €
  • 548-0449A
    Shell vials without plug
    Shell vials without plug
    2,00 ml
    Flat bottom
    12×31 mm
    Borosilicate glass 5.1
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 353,00 €
    353,00 €
    1 * 1.000 items
       353,00 €
  • 548-0048A
    Shell vials, 15 mm PE plug
    Shell vials, 15 mm PE plug
    4,00 ml
    Flat bottom
    15×45 mm
    Borosilicate glass 5.1
    Clear
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 528,00 €
    528,00 €
    1 * 1.000 items
       528,00 €
  • 548-0049A
    Shell vials, 15 mm PE plug
    Shell vials, 15 mm PE plug
    4,00 ml
    Flat bottom
    15×45 mm
    Borosilicate glass 5.1
    Amber
    Yes
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 849,00 €
    849,00 €
    1 * 1.000 items
       849,00 €
Inserts
Product Information
  • Catalog #
    Description
    Capacity
    Base
    Ø×H
    Material
    Colour
    Cap included
    Availability
    Price
  • Micro-insert, 13 mm top
    0,30 ml
    Conical bottom
    6×43,5 mm
    Borosilicate glass 5.1
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 382,00 €
    382,00 €
    1 * 1.000 items
       382,00 €
Product Information
  • Catalog #
    Description
    Capacity
    Base
    Ø×H
    Material
    Colour
    Cap included
    Availability
    Price
  • Micro-insert, 13 mm top
    0,10 ml
    Conical bottom
    5×34 mm
    Borosilicate glass 5.1
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 420,00 €
    420,00 €
    1 * 1.000 items
       420,00 €
Plugs
Product Information
  • Catalog #
    Description
    Cap colour
    Cap Size
    Availability
    Price
  • 8 mm PE-plug with insertion barrier
    Transparent
    8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 101,00 €
    101,00 €
    1 * 1.000 items
       101,00 €
Product Information
  • Catalog #
    Description
    Cap colour
    Cap Size
    Availability
    Price
  • 8 mm PE-plug
    Transparent
    8 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 65,20 €
    65,20 €
    1 * 1.000 items
       65,20 €
Product Information
  • Catalog #
    Description
    Cap colour
    Cap Size
    Availability
    Price
  • 15 mm PE-plug
    Transparent
    15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 222,00 €
    222,00 €
    1 * 1.000 items
       222,00 €