Orders LinkContactUsLinkComponent
Label printer, M210-LAB
Label printer, M210-LAB
Supplier:   Brady
Label printer, M210-LAB
Supplier:  Brady
Certificates
Label printer, M210-LAB
2 Options Available Below
Product Details & Documents
The M210-LAB portable label printer is 'an all in one' labeller for research, academic and clinical labs. With label materials designed specifically for harsh environments, the M210-LAB printer can quickly and easily create legible labels for flat, curved or highly textured surfaces that stay stuck for years, even when exposed to extreme temperatures and chemicals.
  • Monocolour
  • Engineered long lasting material quality
  • Prints durable 'true sized' 6 to 19 mm wide identification to maximise amount of data
  • Adjusts the length of the labels using high performance continuous material
  • Ergonomic cutter and label grabber, holds the label after cutting
  • Easy to use: ABC keypad, graphics library, hot keys and menu functions for fast label creation
  • Smart Cell technology for automatic label set-up and automatic formatting
  • 2 year warranty
Languages available include Croatian; Czech; Danish; Dutch; English (US); Estonian; Finnish; French; German; Hungarian; Italian; Norwegian (Bokmål); Polish; Portuguese; Romanian; Slovak; Slovenian; Spanish (universal); Swedish; Turkish.
Accessories information: Optional multifunctional accessory consists of magnet, flashlight, stand, belt loop and pole strap. 2 AAA batteries required (not included).
Delivery information: The 'carton pack' model ships in a carton pack with a quick start guide and an M21-750-7425 label cartridge.
The kits ship in a hard case with a M210-Lab printer, a Li-ion battery, an EU or UK power cord, a quick start guide, and a M21-750-7425 label cartridge.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • BRDY152261
  • BRDY311319
  • Label:
  • Continuous and die-cut
  • Continuous and die-cut
  • Print resolution:
  • 203 dpi
  • 203 dpi
Specifications
Product Family Options
Product Information
  • Print width
    W×D×H
    Availability
    Price
  • BRDY152261
    M210-LAB Label printer, carton pack, no plug
    M210-LAB Label printer, carton pack, no plug
    16,00 mm
    114,30×63,50×241,30 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 110,00 €
    110,00 €
    1 * 1 items
       110,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BRDY152261
    Label
    Continuous and die-cut
    Print resolution
    203 dpi
  • BRDY311319
    M210-LAB Label printer kit, EU plug
    M210-LAB Label printer kit, EU plug
    16,00 mm
    114,30×63,50×241,30 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 175,00 €
    175,00 €
    1 * 1 Each
       175,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BRDY311319
    Label
    Continuous and die-cut
    Print resolution
    203 dpi
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Belt clip for M210 and M211 label printers
    Belt clip for M210 and M211 label printers
    BRDY170385
    Log in for availability
    1 * 1 items - 11,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Hard case for M210 and M210-LAB label printers
    Hard case for M210 and M210-LAB label printers
    BRDY170421
    Log in for availability
    1 * 1 items - 44,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Multifunctional accessory for M210 and M210-LAB label printers
    Multifunctional accessory for M210 and M210-LAB label printers
    BRDY170424
    Log in for availability
    1 * 1 items - 35,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Magnet only accessory for M210 and M210-LAB label printers
    Magnet only accessory for M210 and M210-LAB label printers
    BRDY170425
    Log in for availability
    1 * 1 items - 16,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Brady workstation CLP/GHS app via download
    Brady workstation CLP/GHS app via download
    BRDY195526
    Log in for availability
    1 * 1 items - 369,99 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Workstation lockout writer software app
    Workstation lockout writer software app
    BRDY145445
    BBP®31, BBP®33, BBP®85, i3300
    Log in for availability
    1 * 1 items - 833,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Brady workstation scan and print software suite
    Brady workstation scan and print software suite
    BRDY149445
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    Log in for availability
    1 * 1 items - 833,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Brady workstation automation software suite
    Brady workstation automation software suite
    BRDY149187
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    Log in for availability
    1 * 1 items - 833,00 €