Orders LinkContactUsLinkComponent
AirTeach software bundle for interactive microscopy class
AirTeach software bundle for interactive microscopy class
AirTeach software bundle for interactive microscopy class
Catalog # 630-3433
Supplier:   LEICA MICROSYSTEMS
AirTeach software bundle for interactive microscopy class
Catalog # 630-3433
Supplier:  LEICA MICROSYSTEMS
AirTeach software bundle for interactive microscopy class
AirTeach software bundle for interactive microscopy class
AirTeach software bundle for interactive microscopy class
Price
38,24 €
Log in for availability
1 * 1 items
Product Details & Documents
The AirTeach software bundle allows users to digitally share microscopy images from various microscope sources. Students are more engaged and inspired in an interactive learning environment that makes sharing and comparing of results fun for everyone.
  • AirTeach software empowers teachers
  • See a thumbnail overview with live images of all connected microscopes in class
  • Share up to four images on a large screen projection to allow comparisons
  • Annotate a selected image live in front of the whole class
Use personal devices and stay connected. The Leica AirLab app allows for easy learning and sharing. Capture, measure, annotate, and archive microscope images with a mobile device. Even make a video of the sample in HD quality. The Leica AirLab app is free of charge and easy to use for iOS and Android, as well as PC or MAC options. Computer users can use the Leica Imaging software to connect to the camera and work with the images. For PCs, use Leica Application Suite software, and use Leica Acquire for Mac.
Ordering information: AirTeach software is compartible with the following cameras and microscopes: ICC50W&E, EZ4W&E, S9i, and IC90E.
Delivery information: AirTeach software bundle CD includes: AirTeach PC software for teacher control of image sharing, AirClass PC software for using PC as an image source, camera configuration program for optimising camera settings, IT requirements, and user manual.
Specifications
  • Description:
    AirTeach software bundle for interactive microscopy class