Orders LinkContactUsLinkComponent
Grade 520 a ½ qualitative filter papers, Whatman™
Grade 520 a ½ qualitative filter papers, Whatman™
Certificates
Grade 520 a ½ qualitative filter papers, Whatman™
3 Options Available Below
Product Details & Documents
A thin paper with great wet strength and a very high flow rate. Frequently used in technical applications such as the filtration of viscous liquids and emulsions (e.g., sweetened juices, spirits and syrups, resin solutions, oils, or plant extracts).
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.
Also available in flat stock form as Grade 520 a.
Specifications for Product Family
Specifications
  • Catalog No:
  • WHAT10331494
  • WHAT10331456
  • 515-8066
  • Grade:
  • 520 a ½
  • 520 a ½
  • 520 a ½
  • Weight:
  • 90 g/m²
  • 90 g/m²
  • 90 g/m²
  • Thickness:
  • 300 µm
  • 300 µm
  • 300 µm
  • Particle retention:
  • 15 - 18 µm
  • 15 - 18 µm
  • 15 - 18 µm
  • Filtration speed:
  • 17,5 s (Herzberg)
  • 17,5 s (Herzberg)
  • 17,5 s (Herzberg)
Specifications
Product Family Options
Product Information
  • Grade
    Size
    Availability
    Price
  • 520 a ½
    450 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 231,00 €
    231,00 €
    1 * 100 items
       231,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10331494
    Grade
    520 a ½
    Weight
    90 g/m²
    Thickness
    300 µm
    Particle retention
    15 - 18 µm
    Filtration speed
    17,5 s (Herzberg)
  • 520 a ½
    500 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 312,00 €
    312,00 €
    1 * 100 items
       312,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    WHAT10331456
    Grade
    520 a ½
    Weight
    90 g/m²
    Thickness
    300 µm
    Particle retention
    15 - 18 µm
    Filtration speed
    17,5 s (Herzberg)
  • 520 a ½
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 101,00 €
    101,00 €
    1 * 100 items
       101,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    515-8066
    Grade
    520 a ½
    Weight
    90 g/m²
    Thickness
    300 µm
    Particle retention
    15 - 18 µm
    Filtration speed
    17,5 s (Herzberg)