Aluminium extension handles
129-0282
- Description:Female ACME thread adapter
- Seq:02
- Pk:1
An ultra-lightweight, aluminum handle featuring three telescoping sections that extend to the desired length. Ideal for reaching high walls and ceilings, inside and around fixtures, and a range of cleanroom support applications.
- Three telescoping sections for custom lengths
- Ideal for extended reach in a range of cleanroom support applications
- Available with extension from 6' to 18' (2 to 5,5 m)
- Fits most mop frames and cleaning tools with threaded attachment sites
The Micronova Aluminium Extendible Mop Handles feature an ACME threaded end which fits Micronova ACME threaded mop frames, brushes, and cleaning tools. Use the SSA-2 'female' Acme Thread Adapter to attach to Micronova’s Quick Disconnect series mops and tools.